Learn More
CD19 Mouse anti-Human, BB700, Clone: SJ25C1 (also known as SJ25-C1), BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566397
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15825019
  • 306.00€ / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD19
Monoclonal
Brilliant Blue 700 (BB700)
25 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Flow Cytometry
SJ25C1 (also known as SJ25-C1)