missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse anti-Human, BB700, Clone: SJ25C1 (also known as SJ25-C1), BD Horizon™
Mouse Monoclonal Antibody
306.00€ - 690.00€
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25C1 (also known as SJ25-C1) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15825019
|
BD Biosciences
566397 |
25 Tests |
306.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15815019
|
BD Biosciences
566396 |
100 Tests |
690.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Human | |
Primary |
SJ25C1 (also known as SJ25-C1) | |
Monoclonal | |
Purified | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |