missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD185 (CXCR5) Rat anti-Mouse, BB700, Clone: 2G8, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566492
Additional Details : Weight : 0.00840kg
Description
CD185 (CXCR5) Monoclonal antibody specifically detects CD185 (CXCR5) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD185 (CXCR5) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
2G8 | |
Brilliant Blue 700 (BB700) | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |