missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, BB700, Clone: MφP9 (also known as MφP-9), BD Horizon™
Mouse Monoclonal Antibody
351.00€ - 730.00€
Specifications
Antigen | CD14 |
---|---|
Clone | MφP9 (also known as MφP-9) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15825079
|
BD Biosciences
566466 |
25 Tests |
351.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15815079
|
BD Biosciences
566465 |
100 Tests |
730.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Human | |
Primary |
MφP9 (also known as MφP-9) | |
Monoclonal | |
Purified | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |