Learn More
CD14 Mouse anti-Human, Alexa Fluor 488, Clone: M5E2, BD
Mouse Monoclonal Antibody
318.00€ - 427.00€
Specifications
Antigen | CD14 |
---|---|
Clone | M5E2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15811259
|
BD Biosciences
561706 |
25 Tests |
318.00€
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15838998
|
BD Biosciences
557700 |
100 Tests |
427.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD14 | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
M5E2 | |
Monoclonal | |
Mouse | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
Human CD14 Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.