missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse anti-Human, Rhesus, Alexa Fluor™ 647, Clone: 3.9, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565912
Additional Details : Weight : 0.00840kg
Description
CD11c Monoclonal antibody specifically detects CD11c in Human,Rhesus samples. It is validated for Flow Cytometry.Specifications
CD11c | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
CD11c | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
3.9 | |
Alexa Fluor 647 | |
0.1 mg | |
Primary | |
Human, Rhesus Monkey | |
IgG1 κ |