missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CASP10 (Human) Recombinant Protein

Human CASP10 partial ORF ( NP_116756.2, 19 a.a. - 187 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000843-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16013725

  • 341.70€ / 10µg

Estimated Shipment: 11-01-2022
Add to basket

Description

Description

  • Sequence: SFREKLLIIDSNLGVQDVENLKFLCIGLVPNKKLEKSSSASDVFEHLLAEDLLSEEDPFFLAELLYIIRQKKLLQHLNCTKEEVERLLPTRQRVSLFRNLLYELSEGIDSENLKDMIFLLKDSLPKTEMTSLSFLAFLEKQGKIDEDNLTCLEDLCKTVVPKLLRNIEK
Specifications

Specifications

NP_116756.2
caspase 10, apoptosis-related cysteine peptidase
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CASP10
GST
843
Wheat germ expression system
10 μg
ALPS2, FLICE2, MCH4
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers