missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CARHSP1 (Human) Recombinant Protein
Human CARHSP1 full-length ORF ( NP_001035941.1, 1 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | NP_001035941.1 |
---|---|
Gene ID (Entrez) | 23589 |
Name | calcium regulated heat stable protein 1, 24kDa |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127502
|
Abnova™
H00023589-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16137502
|
Abnova™
H00023589-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MSSEPPPPPQPPTHQASVGLLDTPRSRERSPSPLRGNVVPSPLPTRRTRTFSATVRASQGPVYKGVCKCFCRSKGHGFITPADGGPDIFLHISDVEGEYVPVEGDEVTYKMCSIPPKNEKLQAVEVVITHLAPGTKHETWSGHVISS
Specifications
NP_001035941.1 | |
calcium regulated heat stable protein 1, 24kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
CRHSP-24, CSDC1, MGC111446 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
23589 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARHSP1 | |
GST |