Learn More
Erlab™ Captair Smart Filtering Fume Hood Mobicap
Erlab™ Captair Smart Filtering Fume Hood Mobicap offers a mobile, rolling bench for Captair Smart fume hoods.
863.90€ - 1314.40€
Specifications
Height (Metric) | 890 mm |
---|---|
Includes | Without Work Surface |
Product Type | Mobile Rolling Cart Work Bench |
Material | Metal |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15515184
|
Erlab™
I0560000201 |
Smart 321 Fume Hood |
863.90€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15594510
|
Erlab™
I0560000201 |
Smart 321 Fume Hood |
863.90€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15535184
|
Erlab™
I0561000201 |
Smart 391 Fume Hood |
1097.10€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15575184
|
Erlab™
I0563000201 |
Smart 392 Fume Hood |
1208.40€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15555184
|
Erlab™
I0562000201 |
Smart 481 Fume Hood |
1314.40€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
- For Captair Smart filtering fume hoods
- Equipped with 4 wheels and 2 self-locks
- Allows the fume hood to be safely moved
Specifications
890 mm | |
Without Work Surface | |
Mobile Rolling Cart Work Bench | |
Metal |