Description
- For a wide variety of fume hoods and cabinets
- Filters for organic vapors, acid vapors and more
Specifications
Specifications
| Filter Type | BE |
| For Use With (Equipment) | Filtair 834, Filtair XL 1044, Filtair XL 1344 |
| Height (Metric) | 106 mm |
| Depth (Metric) | 405 mm |
| Product Type | Filter |
| Model | MF4BE |
| Width (Metric) Exterior | 755 mm |
| For Use With (Application) | Organic and Acid Vapors |