missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, ≥95.0%, ACS Reagent, Honeywell Fluka™

Catalog Number 15622430
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1305-62-0
CaH2O2
74.09
MFCD00010901
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
6093208
CHEBI:31341
calcium;dihydroxide
[OH-].[OH-].[Ca++]
This item is not returnable. View return policy
1305-62-0
CaH2O2
74.09
MFCD00010901
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
6093208
CHEBI:31341
calcium;dihydroxide
[OH-].[OH-].[Ca++]

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 450°C
Color White
Quantity 2.5 kg
Linear Formula Ca(OH)2
UN Number NONH for all modes of transport
Formula Weight 74.09g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium Hydroxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'