Learn More
Merck™ Calbiochem™ PI-103
A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR.
Brand:  Merck™ 528100-5MG
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 15505225
  • 649.44€ / 5mg
Estimated Shipment date
from Supplier 19-11-2018
Add to basket
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
97% by HPLC
Protect from Light
DNA-PK, PI3-K, and mTOR
C19H16N4O3
DMSO
White solid
5mg
371935-74-9
348.4g/mol
Videos