missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Sulfate, ACS Reagent, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 1000°C |
Color | Colorless |
Quantity | 100 g |
Linear Formula | CdSO4 |
UN Number | UN2570 |
Formula Weight | 208.47g/mol |
Percent Purity | ≥99% |
Loss on Drying | ≤1.0% |
Packaging | Poly Bottle |
Appearance | Crystalline |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'