Learn More
IBA Solutions For Life Science C8-Alkyne-dUpA-PH Transcription Starter
A DNA/RNA oligonucleotide for 5´-labeled transcripts that allows click reaction with a number of azide labels.
Brand:  IBA Solutions For Life Science 5-0710-532
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15623708
  • 2205.00€ /
Estimated Shipment date
from Supplier 28-05-2019
Add to basket
Description
Description
One of the main reasons for the success of IBA in custom oligonucleotide and dinucleotide synthesis is the in-house nucleic acid chemistry facility that enabless production of top quality modified oligonucleotides at competitive prices.
  • Modified dinucleotides for 5' transcript labeling: Transcription starters
  • 5'-Modified ApG can be used in in vitro transcription experiments—it is incorporated only once at the 5'-end
  • Easily introduce fluorescent markers to make transcripts visible
  • Introduce non-radioactive markers like biotin or digoxigenin to isolate transcription products
  • In principle, all of the available dyes and non-radioactive reporter groups can be introduced
Specifications
Specifications
C8-Alkyne-dUpA-PH Transcription Starter
Transcription starter for 5' transcript labeling
RNA oligonucleotide
10 μmol
Videos