missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ C500 M Foam Cut Protection Gloves

Catalog Number 17210431
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Bamboo TwinFlex™ Technology provides level C cut protection, while SoftGrip foam coating on the palm and fingertips enhances grip capabilities.

  • SoftGrip coating on the palm and fingertips for a high level of abrasion resistance
  • Bamboo TwinFlex™ Technology provides level C cut protection
  • Certified in accordance with EN 407 for contact heat up to 100°C
  • Enhanced grip in dry, or slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with Oeko-Tex™ Standard 100
  • Silicone-free according to imprint test
Certifications/Compliance (Europe) EN 407: Contact Heat Level 1
Applications Industrial
Coating Material SoftGrip Foam
Color Green, Black
Material Bamboo, Polyamide, Glass
Size 7
Temperature (Metric) Max 100°C
EN388 Standard 4X4C2
For Use With (Application) Automotive, Construction, Glass, Maintanence, and Metal Work
Gender Unisex
Glove Cuff Style Knit
Series C500 M Foam
Yarn Gauge 15
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'