missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffer Solutions for NucleoMag™ Virus
Additional buffer solutions (e.g., lysis, binding, wash, carrier RNA) for use in magnetic bead-based isolation of viral RNA and DNA from serum and plasma using the NucleoMag™ Virus kit.
Brand: Macherey-Nagel™ 744869.300
Additional Details : Weight : 0.30000kg
Description
NucleoMag™ Virus Kits
- One-tube procedure minimizes risk of cross-contamination
- Small elution volumes > 50 μL possible
- Yield does not depend on elution volume
- Suitable for manual and automated processing
- Proven 96-well format for high throughput
- Proteinase K and Carrier RNA included
- Extraction of viral DNA and RNA from serum or plasma
- Veterinary testing applications
- Norovirus RNA isolation from stool samples
Specifications
Colorless | |
NucleoMag™ Virus kit | |
300 mL | |
Room Temperature (18°C to 25°C) | |
Isolation of viral RNA and DNA from serum or plasma |
Ethanol (55 - < 75%) | |
Ethanol (55 - < 75%) | |
Alcoholic | |
Liquid | |
Wash buffer MV4 |