missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 10.0 (20°C), Violet colored, borax / sodium hydroxide, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
10 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
pH | 10.0 |
Quantity | 1 L |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Buffer solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'