Learn More
Fisherbrand™ Buffer Saver Blocks
Physically reduce volume in positive and negative compartments of gel chamber thus reducing buffer requirements.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Economic low gel and buffer volumes
- Available for multiSUB Horizontal Gel Units
Specifications
Sub-Gel Mini Horizontal Gel Electrophoresis Unit | |
Buffer Saver Block | |
2 Each |