missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Reagent Bottles with Screw Caps

Catalog Number p-4380260
Click to view available options
Capacity (Metric):
10,000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

VITLAB polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • Good chemical resistance for long-term storage of liquids.
  • Screw caps help protect against outside contamination and leakage.
  • Autoclavable at 121 C, according to DIN EN 285
Capacity 338.1 oz., 10000mL
Closure Material Polypropylene
Closure Size GL 63
Closure Type Screw Cap
Diameter (Metric) 222 mm
Height (English) 15.51 in.
Height (Metric) 394 mm
Capacity (Metric) 10,000 mL
Capacity (English) 338.1 oz.
Diameter (English) 8.74 in.
Material Polypropylene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'