missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Borosilicate Glass Type 151 D Filter Funnels

Made of borosilicate glass. DURAN™Borosilicate Glass Type 151 D Filter Funnels are used for qualitative inorganic analysis and preparative chemistry.

449.00€ - 525.00€

Specifications

Capacity (Metric) 4000 mL
Material Borosilicate Glass
Diameter (Metric) Disc 175mm
Product Type Filter funnel
View More Specs

Products 4
Product Code Brand Porosity Price Quantity & Availability  
Product Code Brand Porosity Price Quantity & Availability  
10291931 View Documents Duran™
258526104
1 460.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
 
10068090 View Documents Duran™
258526207
2 460.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
 
10508291 View Documents Duran™
258526301
3 449.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
 
10614481 View Documents Duran™
258526404
4 525.00€
Each View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
 
Description

Description

  • Good thermal shock and chemical resistance 
  • Mate to the filtering flask via a conical rubber seal (GUKO)
Specifications

Specifications

4000 mL
Borosilicate Glass
175mm
Filter funnel