missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Borosilicate Glass Specimen Jar

Brand:  Hirschmann™ 242093804

Additional Details : Weight : 0.80000kg

Product Code. 10384081

  • 62.22€ / Pack of 12


Estimated Shipment
15-04-2020
Add to basket

Specifications

Specifications

Borosilicate Glass
110mm
Ground-in Knobbed Lid
Specimen Jar
175 mL
203mm
12Pack
Videos
Documents