Learn More
Duran™ Borosilicate Glass Filter Crucibles
146.00€ - 515.00€
| Product Code | Brand | Porosity | Capacity (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Porosity | Capacity (Metric) | Price | Quantity & Availability | ||||
| 11776848 |
Duran™
258512204 |
2 | 30 mL |
151.00€ Pack of 10 |
|
11756273 |
Duran™
258511105 |
1 | 15 mL |
146.00€ Pack of 10 |
|
11757188 |
Duran™
258511208 |
2 | 15 mL |
146.00€ Pack of 10 |
|
11706293 |
Duran™
258512101 |
1 | 30 mL |
166.00€ Pack of 10 |
|
11736293 |
Duran™
258513106 |
1 | 50 mL |
220.00€ Pack of 10 |
|
11732095 |
Duran™
258513209 |
2 | 50 mL |
220.00€ Pack of 10 |
|
11776248 |
Duran™
258510203 |
2 | 8 mL |
146.00€ Pack of 10 |
|
11786283 |
Duran™
258511302 |
3 | 15 mL |
146.00€ Pack of 10 |
|
11716293 |
Duran™
258512307 |
3 | 30 mL |
151.00€ Pack of 10 |
|
11746303 |
Duran™
258513303 |
3 | 50 mL |
220.00€ Pack of 10 |
|
11796283 |
Duran™
258511405 |
4 | 15 mL |
175.00€ Pack of 10 |
|
11736273 |
Duran™
258510306 |
3 | 8 mL |
146.00€ Pack of 10 |
|
11767188 |
Duran™
258512401 |
4 | 30 mL |
168.00€ Pack of 10 |
|
11756303 |
Duran™
258513406 |
4 | 50 mL |
254.00€ Pack of 10 |
|
11746273 |
Duran™
258510409 |
4 | 8 mL |
166.00€ Pack of 10 |
|
11726293 |
Duran™
258512504 |
5 | 30 mL |
327.00€ Pack of 10 |
|
11777188 |
Duran™
258513509 |
5 | 50 mL |
515.00€ Pack of 10 |
|
Description
- Good thermal shock and chemical resistance
- Typical applications: qualitative inorganic analysis and preparative chemistry
Specifications
| Filter Crucible | |
| Borosilicate Glass |