Learn More
Duran™ Borosilicate Glass Filter Crucibles
146.00€ - 515.00€
Specifications
Product Type Filter Crucible
Material Borosilicate Glass
View More Specs
Products 17
Product Code Brand Porosity Capacity (Metric) Price Quantity & Availability  
Product Code Brand Porosity Capacity (Metric) Price Quantity & Availability  
11776848 View Documents Duran™
258512204
2 30 mL 151.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11756273 View Documents Duran™
258511105
1 15 mL 146.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11757188 View Documents Duran™
258511208
2 15 mL 146.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11706293 View Documents Duran™
258512101
1 30 mL 166.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11736293 View Documents Duran™
258513106
1 50 mL 220.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
11732095 View Documents Duran™
258513209
2 50 mL 220.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
11776248 View Documents Duran™
258510203
2 8 mL 146.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11786283 View Documents Duran™
258511302
3 15 mL 146.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11716293 View Documents Duran™
258512307
3 30 mL 151.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11746303 View Documents Duran™
258513303
3 50 mL 220.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
11796283 View Documents Duran™
258511405
4 15 mL 175.00€
Pack of 10 View alternative products
Save Up to 
»
In Stock 2
Add to basket
11736273 View Documents Duran™
258510306
3 8 mL 146.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11767188 View Documents Duran™
258512401
4 30 mL 168.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
11756303 View Documents Duran™
258513406
4 50 mL 254.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
01-07-2020
Add to basket
11746273 View Documents Duran™
258510409
4 8 mL 166.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11726293 View Documents Duran™
258512504
5 30 mL 327.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
11777188 View Documents Duran™
258513509
5 50 mL 515.00€
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
03-07-2020
Add to basket
Description
Description
  • Good thermal shock and chemical resistance
  • Typical applications: qualitative inorganic analysis and preparative chemistry
Specifications
Specifications
Filter Crucible
Borosilicate Glass