missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Extra Pure, SLR, Eur. Ph., Fisher Chemical™

Catalog Number p-7072866
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
OB(O)O
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
OB(O)O

Boric Acid, Extra Pure, SLR, Eur. Ph.

TRUSTED_SUSTAINABILITY
pH 3.8 to 4.8
Color White
Melting Point 169°C
Packaging Plastic powder jar
Vapor Pressure 2.7mbar at 20°C
Quantity 500 g
Physical Form Powder Solid
Formula Weight 61.83g/mol
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'