missing translation for 'onlineSavingsMsg'
Learn More

BOC-2-piperidylmethanol, 97%, Thermo Scientific™

Catalog Number p-4258786
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
157634-00-9
C11H21NO3
215.29
PZTAGFCBNDBBFZ-UHFFFAOYSA-N
n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
2763853
tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCCC1CO
This item is not returnable. View return policy
157634-00-9
C11H21NO3
215.29
PZTAGFCBNDBBFZ-UHFFFAOYSA-N
n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
2763853
tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCCC1CO
Melting Point 74.4°C to 75.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material BOC-2-Piperidylmethanol
compliance-icons
Product Identifier
  • BOC-2-Piperidylmethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'