Learn More
Kimberly-Clark™ Scott™ XTRA Blue Interfolded Hand Towel
Click to view available options
Dimensions (L x W):
315 x 200 mm
Unit Size:
Pack of 2700
Specifications
Specifications
| Product Type | Scott Xtra Interfolded Hand Towel |
| Length (Metric) | 315 mm |
| Width (Metric) | 200 mm |
| Material | AIRFLEX™ Fabric |
| Certifications/Compliance | ISO 14001, ISO 9001 |
| Color | Blue |
| Dimensions (L x W) | 315 x 200 mm |
| Format | Interfold |
| For Use With (Application) | For food contact areas and busy office and hotel washrooms |
| For Use With (Equipment) | Aquarius Folded Hand Towel Dispenser |
| Show More |