Learn More
Uvex Black/Silver S2 69788 Motion Style Low Shoe - Width 11
Uvex Motion Style safety shoes are lightweight with optimizes wearer comfort using Climazone climate management
Brand:  Uvex 6978849
Additional Details : Weight : 1.87400kg
 View more versions of this product
Product Code. 17214331
  • 140.00€ / 1 pair
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
Uvex Motion Style Shoe 6978 S2 SRC
  • Extremely light, fashionable shoe
  • Breathable, water-resistant high-tech microvelour
  • Ultra breathable distance mesh lining
  • Made from synthetic textiles, suitable for people allergic to chrome
  • Elastic laces for quick, individual adjustment
  • Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Soft padded collar
  • Comfortable removable insole, antistatic with moisture control
  • Uvex 3D Hydroflex™ foam insole for exceptional shock absorption in heel and under front of foot
  • Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
  • PUR outsole
  • Article No. 6978.8
  • Standard EN ISO 20345:2011 S2 SRC
  • Sizes: 36 to 50
Specifications
Specifications
Black, Silver
49
Steel
EN ISO 20345:2011
Standard width
Yes
S2
Microvelour
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex Motion Classic
No
SRC