Learn More
Fisherbrand™ Black Phenolic Caps
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
Description
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Closure Liner | Polyethylene Cone |
Closure Material | Phenolic |
Color | Black |
Quantity | 1600 |
Closure Color | Black |
Neck Finish | 38-400 |
Closure Included | No |