Learn More
Bisphenol B 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30313807
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30313807
Brand: Dr. Ehrenstorfer DREA10655670ME100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mL
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol