missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Mouse VEGF164 Protein, His, Avitag™

Biotinylated Protein

457.25€ - 2011.89€

Specifications

Molecular Weight (g/mol) 22.5 kDa
Source Biotinylated Mouse VEGF164, His,Avitag (VE4-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Accession # Q00731-2).
Immunogen Ala 27 - Arg 190
Storage Requirements -20°C
Gene Alias RP1-261G23.1,MGC70609,MVCD1,VEGFA,VPF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15827927
View Documents Promotion Details
ACROBiosystems
VE4-M82Q3-25UG
25 μg
457.25€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
15817927
View Documents Promotion Details
ACROBiosystems
VE4-M82Q3-200UG
200 μg
2011.89€
200µg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
Description

Description

Vascular endothelial growth factor A (VEGFA) is also known as Vascular permeability factor (VPF). VEGFA belongs to the PDGF/VEGF growth factor family. VEGFA is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. VEGFA is produced by a group of three major isoforms as a result of alternative splicing and if any three isoforms are produced (VEGFA120, VEGFA164, and VEGFA188) then this will not result in vessel defects and death of the full VEGFA knockout in mice.

This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 22.5 kDa. As a result of different glycosylation, the protein migrates as 28-33 kDa under reducing (R) condition, and 46-60 kD
Specifications

Specifications

22.5 kDa
Ala 27 - Arg 190
RP1-261G23.1,MGC70609,MVCD1,VEGFA,VPF
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Biotinylated Mouse VEGF164, His,Avitag (VE4-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Accession # Q00731-2).
-20°C
VEGF164
Mouse
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
Videos
SDS
Documents

Documents

Product Certifications