missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human SIRP alpha / CD172a Protein, Fc,Avitag™

MALS verified

Brand:  ACROBiosystems CDA-H82F2-25UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15816357

  • 353.08€ / 25µg


Estimated Shipment: 22-10-2021
Add to basket

Description

Description

Tyrosine-protein phosphatase non-receptor type substrate 1 (SHPS1) is also known as CD172 antigen-like family member A (CD172a), Macrophage fusion receptor, MyD-1 antigen, Signal-regulatory protein alpha (SIRPA or SIRP alpha) or p84, is a member of the SIRP family, and also belongs to the immunoglobulin superfamily. SIRP alpha is Ubiquitous and highly expressed in brain. SIRPA / CD172a is immunoglobulin-like cell surface receptor for CD47 and acts as docking protein and induces translocation of PTPN6, PTPN11 and other binding partners from the cytosol to the plasma membrane. SIRPA / SHPS-1 supports adhesion of cerebellar neurons, neurite outgrowth and glial cell attachment and may play a key role in intracellular signaling during synaptogenesis and in synaptic function By similarity. SIRPA / MyD1 involved in the negative regulation of receptor tyrosine kinase-coupled cellular responses induced by cell adhesion, growth factors or insulin and mediates negative regulation of phagocytosis, mast cell activation and dendritic cell activation. CD47 binding prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 65.8 kDa. The protein migrates as 75-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_001035111.1
HEK293
Human
65.8 kDa
25 µg
Glu 31 - Arg 370
SHPS1,SIRPA,CD172A,BIT,MFR,MYD1,P84,PTPNS1
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human SIRP alpha, Fc,Avitag (CDA-H82F2) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession # P78324-1).
-20°C
SIRP alpha
Fc Tag and Avi Tag
Videos
SDS
Documents
Certificates