Learn More
ACROBiosystems Biotinylated Human PD-L2 / B7-DC Protein, Avitag™,His Tag
Recommended for biopanning
453.47€ - 1995.26€
Specifications
Accession Number | AAI13679 |
---|---|
Molecular Weight (g/mol) | 25.2 kDa |
Source | Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # AAI13679). |
Immunogen | Leu 20 - Pro 219 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15807517
|
ACROBiosystems
PD2-H82E8-25UG |
25 μg |
453.47€
25µg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
15897507
|
ACROBiosystems
PD2-H82E8-200UG |
200 μg |
1995.26€
200µg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
Description
Programmed cell death 1 ligand 2 (PD-L2 or PDCD1 ligand 2) is also known as Butyrophilin B7-DC, CD antigen CD273, which belongs to the immunoglobulin superfamily or BTN/MOG family. The expression of PD-L2 is up-regulated by IFNG/IFN-gamma stimulation in monocytes and induced on dendritic cells grown from peripheral blood mononuclear cells with CSF2 and IL-4. PD-L2 Involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner. PD-L2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.2 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAI13679 | |
Biotinylated Human PD-L2, Avitag,His Tag (recommended for biopanning) (PD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # AAI13679). | |
-20°C | |
PD-L2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
25.2 kDa | |
Leu 20 - Pro 219 | |
PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |