Learn More
ACROBiosystems Biotinylated Human PD-L1 / B7-H1 Protein, Fc,Avitag™
MALS verified
457.25€ - 2011.89€
Specifications
Accession Number | NP 054862.1 |
---|---|
Molecular Weight (g/mol) | 53.6 kDa |
Source | Biotinylated Human PD-L1, Fc,Avitag (PDL-H82F2) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1). |
Immunogen | Phe 19- Arg 238 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15807537
|
ACROBiosystems
PDL-H82F2-25UG |
25 μg |
457.25€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15897527
|
ACROBiosystems
PDL-H82F2-200UG |
200 μg |
2011.89€
200µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 53.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP 054862.1 | |
Biotinylated Human PD-L1, Fc,Avitag (PDL-H82F2) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1). | |
-20°C | |
PD-L1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
53.6 kDa | |
Phe 19- Arg 238 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |