missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human LILRB5 / CD85c / LIR-8 Protein, Fc, Avitag™

Biotinylated Protein

Brand:  ACROBiosystems CDC-H82F8-25UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15816377

  • 353.08€ / 25µg


Estimated Shipment: 19-10-2021
Add to basket

Description

Description

Leukocyte immunoglobulin-like receptor subfamily B member 5 (LILRB5), also known as CD85c and LIR-8, belongs to the leukocyte immunoglobulin-like receptor (LIR) family of transmembrane glycoproteins that negatively regulate immune cell activation. Several other LIR subfamily B receptors are expressed on immune cells where they bind to MHC class I molecules on antigen-presenting cells and inhibit stimulation of an immune response.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.8 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_006831.1
HEK293
Human
75.8 kDa
25 µg
Gly 24 - Gly 458
LILRB5,CD85c,LIR-8
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human LILRB5, Fc,Avitag (CDC-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 458 (Accession # O75023-1).
-20°C
LILRB5
Fc Tag and Avi Tag
Videos
SDS
Documents
Certificates