missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human LILRB3 / CD85a / ILT5 Protein, Fc, Avitag™

Biotinylated Protein

Brand:  ACROBiosystems CDA-H82F6-200UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15826357

  • 1392.13€ / 200µg


Estimated Shipment: 19-10-2021
Add to basket

Description

Description

Leukocyte immunoglobulin-like receptor subfamily B member 3 is also known as LILRB3,ILT-5 or CD85a. LILRB3 plays an role as receptor for class I MHC antigens, which activated upon coligation of LILRB3 and immune receptors, such as FCGR2B and the B-cell receptor. LILRB3 and LILRA6 represent a pair of inhibitory/activating receptors with identical extracellular domains and unknown ligands. LILRB3 can mediate inhibitory signaling via immunoreceptor tyrosine-based inhibition motifs in its cytoplasmic tail whereas LILRA6 can signal through association with an activating adaptor molecule, FcRγ.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 74.6 kDa. As a result of glycosylation, the protein migrates as 80-105 kDa under reducing (R) condition, and 150 kDa under non-
Specifications

Specifications

AAI12199
HEK293
Human
74.6 kDa
200 µg
Gly 24 - Glu 443
LILRB3,CD85a,ILT5
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human LILRB3, Fc,Avitag (CDA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 443 (Accession # AAI12199).
-20°C
LILRB3
Fc Tag and Avi Tag
Videos
SDS
Documents
Certificates