missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human LILRA2 / CD85h / ILT1 Protein, His, Avitag™

Biotinylated Protein

Brand:  ACROBiosystems LI2-H82E9-200UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15817257

  • 1392.13€ / 200µg


Estimated Shipment: 19-10-2021
Add to basket

Description

Description

LILRA2 (Leukocyte immunoglobulin like receptor A2), also known as CD85H and LIR7 (Leukocyte immunoglobulin-like receptor 7). It belongs to the subfamily A class of LIR receptors (LILRAs), which are single-pass type I transmembrane proteins with 2-4 extracellular Ig-like domains, a transmembrane domain (TM), and a short cytoplasmic tail. LILRA2 contains 4 Ig-like C2 type domains in the extracellular region. LILRA2 does not bind class I MHC antigens. LILRA2 is expressed predominantly on monocytes and B cells, and at lower levels on dendritic cells and natural killer cells. LILRA2 is an activating receptor that inhibits dendritic cell differentiation and antigen presentation and suppresses innate immune response.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

Q8N149.2
HEK293
Human
50.7 kDa
200 µg
Gly 24 - Asn 449
ILT1,LIR7,CD85H,LIR-7,LILRA2,ILT1CD85H,CD85h,ILT1,ILT-1,ILT1CD85H,CD85h antigen,CD85 antigen-like family member H
Precision Avi
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>90% as determined by SDS-PAGE
Biotinylated Human LILRA2, His,Avitag (LI2-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 449 (Accession # Q8N149-1).
-20°C
LILRA2
His Tag and Avi Tag
Videos
SDS
Documents
Certificates