Learn More
ACROBiosystems Biotinylated Human IL-21 Protein, Fc,Avitag™
MALS verified
Brand:  ACROBiosystems IL1-H82F7-200UG
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15827027
  • 2011.89€ / 200µg
Estimated Shipment: 26-04-2024
Description
Interleukin-21 (IL-21) is a secreted protein which belongs to the IL-15 / IL-21 family. Interleukin-21 / IL-21 belongs to a family of cytokines that bind to a composite receptor consisting of a private receptor (IL21R) and the common cytokine receptor gamma chain (gamma(C)). Interleukin-21 / IL-21 impacts a number of cell types, including CD8+ memory T cells, NK cells and subsets of CD4 memory T cells. The IL-21R is widely distributed on lympho-haematopoietic cells. IL-21 is a pleiotropic cytokine produced by CD4+ T cells in response to antigenic stimulation. Its action generally enhances antigen-specific responses of immune cells. IL-21 promotes the anti-tumor activity of CD8+ T-cells and NK cells. IL-21 exerts its effect through binding to a specific type I cytokine receptor, IL-21R, which also contains the γ chain (γc) found in other cytokine receptors including IL-2, IL-4, IL-7, IL-9 and IL-15. The IL-21/IL-21R interaction triggers a cascade of events which includes activation of the tyrosine kinases JAK1 and JAK3, followed by activation of the transcription factors STAT1 and STAT3.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 43.6 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
NP_001193935.1
200 μg
Gln 30 - Ser 162
IL21,Za11,Interleukin-21
Precision Avi
Fc Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
43.6 kDa
Biotinylated Human IL-21, Fc,Avitag (IL1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Ser 162 (Accession # Q9HBE4-1).
-20°C
IL-21
Human
50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5
HEK293
MABSol™ Biotin Labeled Proteins