Learn More
ACROBiosystems Biotinylated Human IL-17 RA / CD217 Protein, Fc, Avitag™
Biotinylated Protein
457.25€ - 2011.89€
Specifications
Accession Number | NP_055154.3 |
---|---|
Molecular Weight (g/mol) | 61.9 kDa |
Source | Biotinylated Human IL-17 RA, Fc,Avitag (ILA-H82F1) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1). |
Immunogen | Leu 33 - Trp 320 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15817087
|
ACROBiosystems
ILA-H82F1-200UG |
200 μg |
2011.89€
200µg |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
15827087
|
ACROBiosystems
ILA-H82F1-25UG |
25 μg |
457.25€
25µg |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
Interleukin 17 receptor A (IL17RA) is also known as cluster of differentiation w217 (CDw217), is a pro-inflammatory cytokine secreted by activated T-lymphocytes, belong to ubiquitous type I membrane glycoprotein, and binds with low affinity to interleukin 17A (IL17A). IL 17R mRNA exhibits a broad tissue distribution, and has been detected in virtually all cells and tissues tested . IL 17RA associates with IL 17RC to form a signaling receptor complex for IL 17 and IL 17F . Ligand and IL 17RA ligation promotes T cell activation and the production of IL - 6, G-CSF, SCF, and multiple pro-inflammatory chemokines. Defects in IL17RA are the cause of familial candidiasis type 5 (CANDF5).
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 61.9 kDa. As a result of glycosylation, the protein migrates as 80-115 kDa under reducing (R) condition, and 150-200 kDa underSpecifications
NP_055154.3 | |
Biotinylated Human IL-17 RA, Fc,Avitag (ILA-H82F1) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1). | |
-20°C | |
IL-17 RA | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
61.9 kDa | |
Leu 33 - Trp 320 | |
CD217,CDw217,IL-17 RA,IL17R,CANDF5,hIL-17 R | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |