missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human HGF R / c-MET Protein, Avitag™,His Tag

MALS verified

Brand:  ACROBiosystems MET-H82E1-200UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15807307

  • 1392.13€ / 200µg


Estimated Shipment: 20-10-2021
Add to basket

Description

Description

Hepatocyte growth factor receptor (HGFR) is also known as mesenchymal-epithelial transition factor (MET), c-Met, and is a glycosylated receptor tyrosine kinase that plays a central role in epithelial morphogenesis and cancer development. HGFR protein possesses tyrosine-kinase activity. The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits, which are disulfide linked to form the mature receptor. HGFR is normally expressed by cells of epithelial origin, while expression of HGF is restricted to cells of mesenchymal origin. Upon HGF stimulation, HGFR induces several biological responses that collectively give rise to a program known as invasive growth. Abnormal HGFR activation in cancer correlates with poor prognosis, where aberrantly active HGFR triggers tumor growth, formation of new blood vessels (angiogenesis) that supply the tumor with nutrients, and cancer spread to other organs (metastasis). HGFR is deregulated in many types of human malignancies, including cancers of kidney, liver, stomach, breast, and brain. Normally, only stem cells and progenitor cells express HGFR, However, cancer stem cells are thought to hijack the ability of normal stem cells to express HGFR, and thus become the cause of cancer persistence and spread to other sites in the body. Various mutations in the HGFR gene are associated with papillary renal carcinoma. HGFR mediates a complex program known as invasive growth. Activation of HGFR triggers mitogenesis, and morphogenesis.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 104.3 kDa. The protein migrates as 40-45 kDa α subunit and 80 kDa β subunit under reducing (R) condition (SDS-PAGE) due to gly
Specifications

Specifications

AAI30421.1
HEK293
Human
104.3 kDa
200 µg
Glu 25 - Thr 932
MET,AUTS9,HGFR,RCCP2,c-Met
Precision Avi
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>90% as determined by SDS-PAGE
Biotinylated Human HGF R, Avitag,His Tag (MET-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1).
-20°C
HGF R
His Tag and Avi Tag
Videos
SDS
Documents
Certificates