Learn More
ACROBiosystems Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His Tag&Fc,Flag Tag, ultra sensitivity (primary amine labeling)
MALS verified
Brand: ACROBiosystems CDD-H82W0-25UG
Additional Details : Weight : 0.00001kg
Description
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
The subunit CD3E carries a human IgG1 Fc fragment at the C-terminus, followed by a polyhistidine tag. The subunit CD3D carries a human IgG1 Fc fragment at the C-terminus, followed by a flag tag. The protein has a calculated MW of 39.2 kDa (CD3E) and 37.5Specifications
NP_000724.1 (CD3E) and NP_000723.1 (CD3D) | |
25 μg | |
Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) | |
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon | |
UltraLys | |
His Tag and Fc Tag and Flag Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.2 kDa (CD3E), 37.5 kDa (CD3D) | |
MABSol™ Biotinylated Human CD3E&CD3D; Heterodimer Protein, Fc,His Tag&Fc;,Flag Tag, primary amine labeling (CDD-H82W0) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # NP_000724.1 (CD3E) and NP_000723.1 (CD3D)). It is the biotinylated form of Human CD3E&CD3D; Heterodimer Protein, Fc,His Tag&Fc;,Flag Tag (CDD-H52W0). | |
-20°C | |
CD3E and CD3D | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |