Learn More
BioTek™ Filters for ELx808™ Absorbance Microplate Reader
Accessory for use with ELx808 Absorbance Microplate Readers.
Brand:  BioTek™ 3100595
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10024782
  • 648.00€ / Each
Estimated Shipment
18-11-2020
Add to basket
Description
Description
Accessory for use with ELx808 Absorbance Microplate Readers.
Specifications
Specifications
Filter (Optional)
595nm
ELx808 Absorbance Microplate Readers