Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with Glass Olive
Allows for the connection of a condenser to a receiving flask. Lenz Laborglasinstrumente™ Bent Receiver Adapters with Glass Olive can be directly heated and withstand typical laboratory thermal variations.
32.75€ - 70.12€
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11886154
|
Lenz Laborglasinstrumente™
05275002 |
NS 19/26 |
33.50€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11806134
|
Lenz Laborglasinstrumente™
05275001 |
NS 14/23 |
32.75€
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11866204
|
Lenz Laborglasinstrumente™
05275003 |
NS 24/29 |
70.12€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11876194
|
Lenz Laborglasinstrumente™
05275004 |
NS 29/32 |
39.38€
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
Description
- NS socket and NS cone
- 105° angle
- Vacuum connection
- Glass olive with drain tip
- Meets DIN 12594 standards
Specifications
Bent Receiver Adapter with Glass Olive | |
Duran Glass |