Learn More
Bel-Art™ SP Scienceware™ Large Pails
Rugged pails with fitted lids/covers and steel-plated bail handles
Brand: Bel-Art™ F16776-0000
Additional Details : Weight : 0.80000kg
Description
- Translucent white pails allow for easier viewing of fluid level
- Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
- 8-qt./7.6L pail has removable steel-plated support ring and bail handle
- 14-qt./13.2L pail has steel-plated swinging bail handle with grip, no support ring
- Large pails meets US Specification MIL-P36257A
Specifications
Steel plated free swinging handle with grip, snug fitting cover | |
13.2L | |
2.99 gal., 13.2L | |
Polypropylene | |
32cm |
Yes | |
US, MIL-PL36257A | |
White | |
Pail |