missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDCA-2 (CD303) Mouse anti-Human, BV421, Clone: V24-785, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566428
Additional Details : Weight : 0.01000kg
Description
BDCA-2 (CD303) Monoclonal antibody specifically detects BDCA-2 (CD303) in Human samples. It is validated for Flow Cytometry.Specifications
BDCA-2 (CD303) | |
Monoclonal | |
Brilliant Violet 421 | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |
Flow Cytometry | |
V24-785 | |
Mouse | |
RUO | |
Human | |
Purified |