Learn More
BD ELISPOT Sets
Sufficient capture antibody, detection antibody, and streptavidin-horseradish peroxidase for 10 × 96 wells
Brand:  BD 551282
Additional Details : Weight : 0.42000kg
 View more versions of this product
Product Code. 15826598
  • 1760.00€ / Each
Estimated Shipment: 13-10-2021
Add to basket
Includes: Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations
Description
Description
  • High sensitivity
  • High throughput, high content analysis
  • Minimal volume of biological material required
  • Suitable for frozen/thawed biological samples
  • Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis
Specifications
Specifications
IL-2
Human
Human IL-2 ELISPOT set
4°C
Cloning, proliferation assays, flow cytometry or other methods of analysis
Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates
10 x 96 Tests