missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mouse IL-12 p70 Flex Set Standard

Lyophilized Mouse IL-12 p70

Brand:  BD 558475

Additional Details : Weight : 0.01000kg

Product Code. 15879418

  • 127.00€ / Each


Estimated Shipment
26-03-2020
Add to basket

Description

Description

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 25,000pg/ml (when reconstituted in 0.4mL Assay Diluent)
  • Regulatory status: RUO
  • Recombinant

Flow cytometry, controls

Order Info

100 tests.

Specifications

Specifications

Mouse IL-12p70 Flex Standard Set
Flow cytometry
Technical data sheet
25000pg/mL
Murine
0.2mL
Aqueous Buffered Solution containing BSA and ProClin™ 150
Lyophilized
100 Tests
RUO
4°C
Videos
Documents