Learn More
BD BBL™/Difco™ TB Decolorizer
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures
Brand: BD 212517
Additional Details : CAS : 7647-01-0 Weight : 0.80000kg
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
Decolorizer | |
For staining mycobacteria by the Kinyoun (cold) and Ziehl-Neelsen (hot) acid-fast procedures | |
Bottle |
4 x 250mL | |
15° to 30°C | |
Decolorizer |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H371-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P233-Keep container tightly closed.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P405-Store locked up.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol, hydrochloric acid
For in vitro Diagnostic Use.