Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Ultrafilter for use with GenPure, GenPure Pro, GenPure xCAD (UF, UF/UV and UF/UV-TOC units only)
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
Brand:  Thermo Scientific™ 50133980
Product Code. 11367161
  • 1065.00€ / Each
Description
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
  • GenPure™†
  • GenPure™ Pro†
  • GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
  • MicroPure™‡
  • Smart2 Pure™‡
‡UF and UF/UV units only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ultrafilter; For use with GenPure, GenPure Pro, GenPure xCAD (UF, UF/UV and UF/UV-TOC units only)
GenPure, GenPure Pro, GenPure xCAD
Water Purification Systems Ultrafilter