missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AVCS 2mL Certified Clear Vial Kit 100pk

Save time and reduce the risk of contamination using this convenience HRV kit. Each kit includes 100 2ml clear vials with caps.

Brand:  Thermo Scientific™ CERT5000-93

Additional Details : Weight : 0.35000kg

Product Code. 15388046

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »


Description

Description

  • AVCS
  • Clear vial and cap
  • Includes bonded septa
  • Supplied in packs of 100
Specifications

Specifications

Clear
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers