missing translation for 'onlineSavingsMsg'
Learn More

Gibertini Elettronica™ EU-C Atex Technical Balances

Designed for use in hazardous environments with the risk of explosion, these scales are classified as ZONE 2. Gibertini Elettronica™ EU-C Atex Technical Balances are built with method of protection nAL for gas group IIC and temperature class T4 (135°C) according to regulation EEC 94/9.

Brand:  Gibertini Elettronica™ EU-C 2002 X

Additional Details : Weight : 4.50000kg

 View more versions of this product

Product Code. 15503886

  • 1370.00€ / Each


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Description

Description

  • LCD display with small-size decimal digits, for easier and more immediate reading
  • Waterproof and acid resistant membrane keyboard. Easy to use with the ON/OFF, TARE, MODE and PRINT keys (RANGE/PRINT for DR model)
  • Indication of the reached stable weight
  • Bar-graph indicator of dosage and remaining capacity of the balance
  • Parameters configurable by menu: reading in g (grams), lb (pound), oz (ounce), ct (carats), pcs (pieces), % (percentage)
  • Automatic end of range calibration (with external mass supplied on request)
  • Memory of the current weight in case of power failure
  • Enhanced anti-shock system
  • Metallic body with epoxide protection
  • Load cell nearly unbreakable and without transportation problems
  • Data output: I/O RS232 adjustable
  • 1, 2 and 5kg masses in class F1 for calibration and special executions available upon request
Specifications

Specifications

≥ 2 kg
Classified as ZONE 2. Built with method of protect
0.01 g
2.5 sec.
EU-C 2002 X
10 to 40°C
4.5kg
Designed for use in hazardous environments with th
355 x 215 x 120mm
Technical Balance
150mm
±0.02g
±0.02g
Metallic body with epoxide protection
240VAC
150mm
240VAC
Yes
Videos