Learn More
ATAGO™ Master-α Refractometer, ATC, Water Resistant
Hand-held refractometer. The ATAGO Master-α Refractometer is equipped with ATC and water resistant at the same time.
Brand: ATAGO™ 2311
Additional Details : Weight : 0.27000kg
Description
- Equipped with Automatic Temperature Compensation and Water Resistant (IP65)
- Easy to read, easy to hold, and easy to clean with a smooth sample stable and curved edges
- Constructed of metal; for salty and acidic samples, choose model with plastic body (MASTER-Pa)
Specifications
Refractometer | |
Master-α | |
Min. Scale ±0.2% | |
10°C to 30°C |
0.0 to 33.0 Brix | |
±0.2% repeatability: ± 0.1% | |
Brix | |
10°C to 30°C |